2',3'-Dideoxyinosine, 98%
2',3'-Dideoxyinosine, 98%
2',3'-Dideoxyinosine, 98%
Thermo Scientific Chemicals

2',3'-Dideoxyinosine, 98%

CAS: 69655-05-6 | C10H12N4O3 | 236.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
1 g
Catalog number J64833.03
also known as J64833-03
Price (USD)
140.65
Online Exclusive
156.00
Save 15.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
140.65
Online Exclusive
156.00
Save 15.35 (10%)
Each
Specifications
Beilstein3619529
CAS69655-05-6
ChEBICHEBI:490877
Chemical Name or Material2',3'-Dideoxyinosine
ColorWhite
View more
2?,3?-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,3′-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

Solubility
Soluble in DMSO or methanol

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Gurpreet Ahluwalia.; David A. Cooney.; Hiroaki Mitsuya.; Arnold Fridland.; Karl P. Flora.; Zhang Hao.; Maha Dalal.; Samuel Broder.; David G. Johns. Initial studies on the cellular pharmacology of 2',3'-dideoxyinosine, an inhibitor of HIV infectivity. Cell. 1987, 36 (22), 3797-3800.
  2. R Yarchoan.; H Mitsuya.; RV Thomas.; JM Pluda.; NR Hartman.; CF Perno.; KS Marczyk.; JP Allain.; DG Johns.; S Broder. In vivo activity against HIV and favorable toxicity profile of 2',3'-dideoxyinosine. Science. 1989, 245(4916), 412-415 .