5,6-Dichlorobenzimidazole riboside, 98%
5,6-Dichlorobenzimidazole riboside, 98%
5,6-Dichlorobenzimidazole riboside, 98%
Thermo Scientific Chemicals

5,6-Dichlorobenzimidazole riboside, 98%

CAS: 53-85-0 | C12H12Cl2N2O4 | 319.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
250 mg
Catalog number J64870.MC
also known as J64870-MC
Price (USD)
328.65
Online exclusive
365.00
Save 36.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
328.65
Online exclusive
365.00
Save 36.35
Each
Chemical Identifiers
CAS53-85-0
IUPAC Name(2R,3R,4S,5R)-2-(5,6-dichloro-1H-1,3-benzodiazol-1-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC12H12Cl2N2O4
InChI KeyXHSQDZXAVJRBMX-DDHJBXDOSA-N
SMILESOC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=CC(Cl)=C(Cl)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Purity≥97.5% (HPLC)
Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.5,6-Dichlorobenzimidazole riboside is used as an inhibitor of RNA Polymerase II. It is nucleoside analogue and used in research to study the mechanisms of cellular regulation. Further, it inhibits some protein kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.5,6-Dichlorobenzimidazole riboside is used as an inhibitor of RNA Polymerase II. It is nucleoside analogue and used in research to study the mechanisms of cellular regulation. Further, it inhibits some protein kinases.

Solubility
Soluble in DMSO to 75mMSoluble in dimethyl sulfoxide, hot ethanol, dimethyl formamide, (1:2) dimethyl formamide:Phosphate buffer saline, water, methanol and pyridine. Slightly soluble in aqueous buffers.

Notes
Incompatible with heat and oxidizing agents.
RUO – Research Use Only

General References:

  1. Liu, Y.; He, X.; Sui, Y.; Yu, R.; Xu, G. Transcription factor IKZF1 is degraded during the apoptosis of multiple myeloma cells induced by kinase inhibition. FEBS Lett. 2015, 589 (17), 2233-2240.
  2. Bartkowiak, B.; Greenleaf, A. L. Expression, Purification, and Identification of Associated Proteins of the Full-length hCDK12/CyclinK Complex. J. Biol. Chem. 2015, 290 (3), 1786-1795.