gamma-Secretase Inhibitor XX
gamma-Secretase Inhibitor XX
gamma-Secretase Inhibitor XX
gamma-Secretase Inhibitor XX
Thermo Scientific Chemicals

gamma-Secretase Inhibitor XX

 
Have Questions?
Catalog NumberQuantity
J64904.0AM
also known as J64904-0&
3 mg
Catalog number J64904.0AM
also known as J64904-0&
Price (USD)
676.65
Special offer
Online exclusive
Ends: 30-Jun-2026
796.00
Save 119.35 (15%)
Each
Quantity:
3 mg
Request bulk or custom format
Price (USD)
676.65
Special offer
Online exclusive
Ends: 30-Jun-2026
796.00
Save 119.35 (15%)
Each
Specifications
CAS209984-56-5
DescriptionWhite
For Use With (Application)A dipeptidic (c)-secretase inhibitor
FormSolid
Formula Weight463.48g/mol
View more
A dipeptidic (c)-secretase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C