Product Image
Thermo Scientific Chemicals

Z-Leu-Glu(OMe)-His-Asp(OMe)-fluoromethyl ketone trifluoroacetate salt hydrate

 
Have Questions?
Catalog NumberQuantity
J64966.MCR
also known as J64966-M^
1 mg
Catalog number J64966.MCR
also known as J64966-M^
Price (USD)
216.65
Special offer
Online exclusive
Ends: 28-Dec-2026
255.00
Save 38.35
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
216.65
Special offer
Online exclusive
Ends: 28-Dec-2026
255.00
Save 38.35
Each
Specifications
DescriptionOff-white to pale brown
For Use With (Application)A cell-permeable inhibitor of caspase-9, which exhibits competitive and irreversible inhibition
FormPowder
Formula Weight690.71(anhy)
Molecular FormulaC32H43FN6O10.xC2HF3O2.yH2O
View more
A cell-permeable inhibitor of caspase-9, which exhibits competitive and irreversible inhibition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C