9-beta-D-Arabinofuranosyladenine, 99%
9-beta-D-Arabinofuranosyladenine, 99%
9-beta-D-Arabinofuranosyladenine, 99%
Thermo Scientific Chemicals

9-beta-D-Arabinofuranosyladenine, 99%

CAS: 5536-17-4 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J65076.06
also known as J65076-06
Price (USD)
57.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
57.90
Each
Specifications
Beilstein624881
CAS5536-17-4
Chemical Name or Material9-beta-D-Arabinofuranosyladenine
ColorWhite
EINECS Number226-893-9
View more
Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 ?M. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 μM. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.

Solubility
Soluble in DMF (10 mg/ml), 0.5 M HCl (50 mg/ml), DMSO (53 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), and water (3 mg/ml at 25°C).

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Gene D. Morse.; Mark J. Shelton.; Alice M. O Donnell. Comparative Pharmacokinetics of Antiviral Nucleoside Analogues. Clinical Pharmacokinetics. 1993, 24, (2), 101-123.
  2. John A. SecristIII.; Anita T. Shortnacy.; John A. Montgomery. Synthesis and biological evaluations of certain 2-halo-2'-substituted derivatives of 9-.beta.-D-arabinofuranosyladenine. J. Med. Chem. 1988, 31, (2), 405-410.