N(alpha)-Acetyl-L-lysine, 99%
N(alpha)-Acetyl-L-lysine, 99%
N(alpha)-Acetyl-L-lysine, 99%
Thermo Scientific Chemicals

N(alpha)-Acetyl-L-lysine, 99%

CAS: 1946-82-3 | C8H16N2O3 | 188.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J65083.06
also known as J65083-06
Price (USD)
273.65
Special offer
Online exclusive
Ends: 30-Jun-2026
322.00
Save 48.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
273.65
Special offer
Online exclusive
Ends: 30-Jun-2026
322.00
Save 48.35 (15%)
Each
Specifications
CAS1946-82-3
ChEBICHEBI:35704
Chemical Name or MaterialN(alpha)-Acetyl-L-lysine
ColorWhite
EINECS Number217-747-5
View more
Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).

Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml--clear colorless solution)).

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Brian P Hudsona.; Maria A Martinez-Yamout.; Jane Dyson, H.; Peter E Wright. Solution structure and acetyl-lysine binding activity of the GCN5 bromodomain1. Journal of Molecular Biology. 2000, 304, (3), 355-370.
  2. Geratz JD.; Cheng MC. The inhibition of urokinase by aromatic diamidines. Thrombosis et Diathesis Haemorrhagica. 1975, 33, (2), 230-243.