Sucrose, Molecular Biology Grade
Sucrose, Molecular Biology Grade
Sucrose, Molecular Biology Grade
Thermo Scientific Chemicals

Sucrose, Molecular Biology Grade

For protein purification by sucrose density gradients | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number J65148.A1
also known as J65148-A1
Price (USD)
79.65
Online Exclusive
88.50
Save 8.85 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
79.65
Online Exclusive
88.50
Save 8.85 (10%)
Each
Chemical Identifiers
CAS57-50-1
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyCZMRCDWAGMRECN-PWPRYFECNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
DNaseNone detected
RNaseNone detected
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Ruby I. MacDonald. Temperature and ionic effects on the interaction of erythroid spectrin with phosphatidylserine membranes. Biochemistry. 1993, 32, (27),6957-6964.
  2. Henry Daniell. Transformation and Foreign Gene Expression in Plants Mediated by Microprojectile Bombardment. Recombinant Gene Expression Protocols. 1997, 62, 463-489.