Cathepsin B Inhibitor III
Cathepsin B Inhibitor III
Cathepsin B Inhibitor III
Cathepsin B Inhibitor III
Thermo Scientific Chemicals

Cathepsin B Inhibitor III

 
Have Questions?
Catalog NumberQuantity
J65347.LB0
also known as J65347-#0
5 mg
Catalog number J65347.LB0
also known as J65347-#0
Price (USD)
325.65
Special offer
Online exclusive
Ends: 30-Jun-2026
383.00
Save 57.35 (15%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
325.65
Special offer
Online exclusive
Ends: 30-Jun-2026
383.00
Save 57.35 (15%)
Each
Chemical Identifiers
CAS134448-10-5
IUPAC Name(2S)-1-[(2S,3S)-3-methyl-2-{[(3S)-3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl]pyrrolidine-2-carboxylic acid
Molecular FormulaC18H29N3O6
InChI KeyZEZGJKSEBRELAS-KSFNBINOSA-N
SMILESCCCNC(=O)[C@H]1OC1C(=O)N[C@@H]([C@@H](C)CC)C(=O)N1CCC[C@H]1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)96.0% min.
CA-074 is a potent and specific inhibitor of cathepsin B in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C