2'-Amino-2'-deoxyinosine, 98%, Thermo Scientific Chemicals
2'-Amino-2'-deoxyinosine, 98%, Thermo Scientific Chemicals
2'-Amino-2'-deoxyinosine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Amino-2'-deoxyinosine, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J65374.MD
also known as J65374-MD
250 mg
Catalog number J65374.MD
also known as J65374-MD
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
100.00
Save 14.35 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
100.00
Save 14.35 (14%)
Each
Specifications
CAS75763-51-8
Chemical Name or Material2'-Amino-2'-deoxyinosine
ColorWhite to Yellow
Formula Weight267.24
IUPAC Name9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only