5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%
5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%
5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%

Substrate for beta→-D-glucosidase | CAS: 15548-60-4 | C14H15BrClNO6 | 408.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number J65677.03
also known as J65677-03
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Specifications
Beilstein1552603
CAS15548-60-4
ChEBICHEBI:75424
Chemical Name or Material5-Bromo-4-chloro-3-indolyl beta-D-glucoside
ColorWhite
View more
Substrate for beta--D-glucosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only