3'-Deoxythymidine
3'-Deoxythymidine
3'-Deoxythymidine
Thermo Scientific Chemicals

3'-Deoxythymidine

CAS: 3416-05-5 | C10H14N2O4 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number J65765.ME
also known as J65765-ME
Price (USD)
403.65
Online Exclusive
448.00
Save 44.35 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
403.65
Online Exclusive
448.00
Save 44.35 (10%)
Each
Specifications
Beilstein21884
CAS3416-05-5
Chemical Name or Material3'-Deoxythymidine
ColorWhite to Yellow
Formula Weight226.23
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only