N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone, Thermo Scientific Chemicals
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone, Thermo Scientific Chemicals
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
J65812.LB0
also known as J65812-#0
5 mg
Catalog number J65812.LB0
also known as J65812-#0
Price (USD)
489.65
Online Exclusive
544.00
Save 54.35 (10%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
489.65
Online Exclusive
544.00
Save 54.35 (10%)
Each
Specifications
CAS187389-53-3
Chemical Name or MaterialN-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone
ColorWhite
Formula Weight263.26
IUPAC Namemethyl (3S)-5-fluoro-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxopentanoate
View more
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone is used as a novel Boc-D(OMe)-FMK inhibitor synthesized using proprietary technology and irreversibly binds to activated caspases to block apoptosis. It is useful for in vitro and in vivo applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone is used as a novel Boc-D(OMe)-FMK inhibitor synthesized using proprietary technology and irreversibly binds to activated caspases to block apoptosis. It is useful for in vitro
RUO – Research Use Only