2'-Fluoro-2'-deoxyuridine, 97%
2'-Fluoro-2'-deoxyuridine, 97%
2'-Fluoro-2'-deoxyuridine, 97%
Thermo Scientific Chemicals

2'-Fluoro-2'-deoxyuridine, 97%

CAS: 784-71-4 | C9H11FN2O5 | 246.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number J65948.14
also known as J65948-14
Price (USD)
401.65
Special offer
Online exclusive
Ends: 30-Jun-2026
472.00
Save 70.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
401.65
Special offer
Online exclusive
Ends: 30-Jun-2026
472.00
Save 70.35 (15%)
Each
Chemical Identifiers
CAS784-71-4
IUPAC Name1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11FN2O5
InChI KeyUIYWFOZZIZEEKJ-XVFCMESISA-N
SMILESOC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0%
Appearance (Color)White to off-white
2'-Fluoro-2'-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.

Solubility
Soluble in methanol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Charles L. Perrin. Mechanism of hydrogen exchange in amides. J. Am. Chem. Soc. 1974, 96, (17), 5628-5631.
  2. Osamu Shimomura; Ikuyoshi Tomita; Takeshi Endo. Cationic polymerization of glycidyl phenyl ether initiated by various arylsulfoniu∼ salts as new thermal latent cationic initiators. Macromol. Rapid Cammun. 1998, 19 (9), 493-497 .