Thermo Scientific Chemicals
Aurora Kinase/Cdk Inhibitor
Catalog number: J65997.LB0
5 mg, Each
Thermo Scientific Chemicals
Aurora Kinase/Cdk Inhibitor
Catalog number: J65997.LB0
5 mg, Each
Quantity
Catalog number: J65997.LB0
also known as J65997-#0
Price (USD)
Price: 391.00
Your price:
Quantity
-
Description
Aurora Kinase/Cdk Inhibitor is a broad spectrum inhibitor of cyclin-dependent kinases and aurora kinases including CDK1/Cyclin B, CDK2/Cyclin A, CDK2/Cyclin E, Aurora-A and Aurora-B. It as been shown to induce growth suppression and mitotic defects, these results suggest that Aurora Kinase/Cdk Inhibitor could be useful for cell cycle analysis and therapy of various cancers, including Ewing's sarcoma.