Nonyl beta-D-glucopyranoside, 98%
Nonyl beta-D-glucopyranoside, 98%
Nonyl beta-D-glucopyranoside, 98%
Thermo Scientific Chemicals

Nonyl beta-D-glucopyranoside, 98%

CAS: 69984-73-2 | C15H30O6 | 306.399 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J66009.06
also known as J66009-06
Price (USD)
825.65
Online Exclusive
917.00
Save 91.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
825.65
Online Exclusive
917.00
Save 91.35 (10%)
Each
Specifications
Beilstein12168
CAS69984-73-2
Chemical Name or MaterialNonyl beta-D-glucopyranoside
ColorWhite
EINECS Number615-042-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only