[Cys8] Renin Substrate Tetradecapeptide, Rat
Thermo Scientific Chemicals

[Cys8] Renin Substrate Tetradecapeptide, Rat

For cellular and molecular biology applications
Have Questions?
Catalog NumberSubstrate Quantity
J66067.LB0
also known as J66067-#0
5mg
Catalog number J66067.LB0
also known as J66067-#0
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Substrate Quantity:
5mg
Request bulk or custom format
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Specifications
ColorWhite to off-white
Molecular Weight (g/mol)1779.05
Recommended StorageStore at -20°C
Storage Requirements-30° to -10°C
Substrate Quantity5mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only