PCR Mycoplasma detection kit - FAQs
PCR Mycoplasma detection kit - FAQs
View additional product information for PCR Mycoplasma detection kit - FAQs (J66117.AMJ)
1 product FAQs found
What storage conditions should I use for the PCR Mycoplasma detection kit (Cat. No. J66117.AMJ)?
The PCR Mycoplasma detection kit should be stored at -20°C. We recommend that you avoid repeated freeze/thaw cycles of the master mix to retain maximum performance.