Laminarin
Laminarin
Laminarin
Thermo Scientific Chemicals

Laminarin

CAS: 9008-22-4 | C18H32O16 | 504.438 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number J66193.03
also known as J66193-03
Price (USD)
126.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
126.00
Each
Chemical Identifiers
CAS9008-22-4
IUPAC Name(3R,4S,5S,6R)-2-{[(2R,3R,4S,5R)-3,5-dihydroxy-2-(hydroxymethyl)-6-{[(3R,4S,5R,6R)-2,3,5-trihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}oxan-4-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyDBTMGCOVALSLOR-VPNXCSTESA-N
SMILESOC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](CO)OC(O[C@@H]3[C@@H](O)C(O)O[C@H](CO)[C@H]3O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Optical Rotation-40 to -50? (c=0.9, H₂O)
Appearance (Color)White to off-white
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

Solubility
Soluble in dimethyl sulfoxide (slightly, heated). Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Hui Dai; Xiao-Qiang Han; Fang-Yuan Gong; Hongliang Dong; Peng-Fei Tu; and Xiao-Ming Gao. Structure elucidation and immunological function analysis of a novel β-glucan from the fruit bodies of Polyporus umbellatus (Pers.) Fries. Glycobiology, 2012,22(12), 1673-1683.
  2. Park, H.K., et al. Induction of apoptosis by laminarin, regulating the insulin-like growth factor-IR signaling pathways in HT-29 human colon cells. Int. J. Molec. Med, 2012,30(4), 734-738.