CREBtide - FAQs
CREBtide - FAQs
View additional product information for CREBtide - FAQs (J66434.EXD, J66434.MCR)
2 product FAQs found
Is the CREBtide peptide acetylated at the N-terminus?
No, the CREBtide peptide is not acetylated at the N-terminus.
What is the amino acid sequence of the CREBtide peptide?
The amino acid sequence of the CREBtide peptide is: Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg