Sphingosine-1-phosphate
Sphingosine-1-phosphate
Sphingosine-1-phosphate
Thermo Scientific Chemicals

Sphingosine-1-phosphate

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J66459.LB0
also known as J66459-#0
Price (USD)
704.65
Special offer
Online exclusive
Ends: 30-Jun-2026
829.00
Save 124.35 (15%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
704.65
Special offer
Online exclusive
Ends: 30-Jun-2026
829.00
Save 124.35 (15%)
Each
Chemical Identifiers
CAS26993-30-6
IUPAC Name{[(2S,3R,4E)-2-amino-3-hydroxyoctadec-4-en-1-yl]oxy}phosphonic acid
Molecular FormulaC18H38NO5P
InChI KeyDUYSYHSSBDVJSM-KRWOKUGFSA-N
SMILESCCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)White to yellow
Infrared spectrumConforms
Assay from Supplier's CofA≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C