Caspase-7 Substrate I, fluorogenic
Thermo Scientific Chemicals

Caspase-7 Substrate I, fluorogenic

For cellular and molecular biology applications
Have Questions?
Catalog NumberSubstrate Quantity
J66513.EXD
also known as J66513-!D
0.5mg
Catalog number J66513.EXD
also known as J66513-!D
Price (USD)
129.65
Special offer
Online exclusive
Ends: 30-Jun-2026
152.00
Save 22.35 (15%)
Each
Substrate Quantity:
0.5mg
Request bulk or custom format
Price (USD)
129.65
Special offer
Online exclusive
Ends: 30-Jun-2026
152.00
Save 22.35 (15%)
Each
Specifications
ColorWhite to off-white
Molecular Weight (g/mol)1327.31
Recommended StorageStore at -20°C
Storage Requirements-30° to -10°C
Substrate Quantity0.5mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only