D-Galacto-D-mannan
Thermo Scientific Chemicals

D-Galacto-D-mannan

CAS: 11078-30-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number J66610.MC
also known as J66610-MC
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Specifications
CAS11078-30-1
Chemical Name or MaterialD-Galacto-D-mannan
ColorWhite
EINECS Number234-299-6
Formula Weight∽200000
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only