4-Methylumbelliferyl beta-D-glucopyranoside, 99%
4-Methylumbelliferyl beta-D-glucopyranoside, 99%
4-Methylumbelliferyl beta-D-glucopyranoside, 99%
Thermo Scientific Chemicals

4-Methylumbelliferyl beta-D-glucopyranoside, 99%

CAS: 18997-57-4 | C16H18O8 | 338.312 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66630.03
also known as J66630-03
Price (USD)
145.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
145.00
Each
Chemical Identifiers
CAS18997-57-4
IUPAC Name4-methyl-7-{[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC16H18O8
InChI KeyYUDPTGPSBJVHCN-YJUJGKJLSA-N
SMILESCC1=CC(=O)OC2=CC(OC3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.5%
FormPowder
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only