4-Methylphenyl beta-D-thiogalactopyranoside, 98%, Thermo Scientific Chemicals
4-Methylphenyl beta-D-thiogalactopyranoside, 98%, Thermo Scientific Chemicals
4-Methylphenyl beta-D-thiogalactopyranoside, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylphenyl beta-D-thiogalactopyranoside, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J66686.03
also known as J66686-03
1 g
Catalog number J66686.03
also known as J66686-03
Price (USD)
75.90
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
75.90
Each
Chemical Identifiers
CAS28244-98-6
IUPAC Name2-(hydroxymethyl)-6-[(4-methylphenyl)sulfanyl]oxane-3,4,5-triol
Molecular FormulaC13H18O5S
InChI KeyIQCLIQLFPVKINX-UHFFFAOYNA-N
SMILESCC1=CC=C(SC2OC(CO)C(O)C(O)C2O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water, warm methanol, DMSO, or DMF.

Notes
Store at 4°C. Protect from heat. Incompatible with heat and oxidizing agents.
RUO – Research Use Only