2,3,5-Tri-O-benzyl-D-arabinofuranose
2,3,5-Tri-O-benzyl-D-arabinofuranose
2,3,5-Tri-O-benzyl-D-arabinofuranose
Thermo Scientific Chemicals

2,3,5-Tri-O-benzyl-D-arabinofuranose

CAS: 37776-25-3 | C26H28O5 | 420.505 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number J66792.09
also known as J66792-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Specifications
CAS37776-25-3
Chemical Name or Material2,3,5-Tri-O-benzyl-D-arabinofuranose
ColorWhite
Formula Weight420.5
IUPAC Name(3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only