RAF265
RAF265
RAF265
Thermo Scientific Chemicals

RAF265

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J67037.MA
also known as J67037-MA
Price (USD)
523.65
Online Exclusive
582.00
Save 58.35 (10%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
523.65
Online Exclusive
582.00
Save 58.35 (10%)
Each
Specifications
CAS927880-90-8
DescriptionPale yellow to yellow
FormSolid
Formula Weight518.41
Molecular FormulaC24H16F6N6O
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C