Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate, 98%
Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate, 98%
Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate, 98%
Thermo Scientific Chemicals

Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate, 98%

CAS: 132465-11-3 | C19H15NO4, C19H15NO4 | 321.33 g/mol
Have Questions?
Catalog NumberQuantity
J67167.MA
also known as J67167-MA
10 mg
Catalog number J67167.MA
also known as J67167-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
Assay98%
CAS132465-11-3
DescriptionYellow-orange
FormPowder
Formula Weight321.33
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only