Rolipram, 98%
Rolipram, 98%
Rolipram, 98%
Thermo Scientific Chemicals

Rolipram, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J67177.MA
also known as J67177-MA
Price (USD)
116.65
Online exclusive
129.00
Save 12.35
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
116.65
Online exclusive
129.00
Save 12.35
Each
Chemical Identifiers
CAS61413-54-5
IUPAC Name4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
Molecular FormulaC16H21NO3
InChI KeyHJORMJIFDVBMOB-UHFFFAOYNA-N
SMILESCOC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Purity≥97.0% (TLC)
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only