U-69593, 99%
U-69593, 99%
U-69593, 99%
Thermo Scientific Chemicals

U-69593, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J67220.LB0
also known as J67220-#0
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Specifications
Assay99%
CAS96744-75-1
DescriptionWhite
FormSolid
Formula Weight356.51
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C