BAY 11-7085, 99%
BAY 11-7085, 99%
BAY 11-7085, 99%
Thermo Scientific Chemicals

BAY 11-7085, 99%

CAS: 196309-76-9 | C13H15NO2S, C13H15NO2S | 249.33 g/mol
Have Questions?
Catalog NumberQuantity
J67222.MA
also known as J67222-MA
10 mg
Catalog number J67222.MA
also known as J67222-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
Assay99%
CAS196309-76-9
DescriptionWhite
FormSolid
Formula Weight249.33
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only