Acetate Buffer (1 M, pH 5.0, Low Endotoxin)
Acetate Buffer (1 M, pH 5.0, Low Endotoxin)
Acetate Buffer (1 M, pH 5.0, Low Endotoxin)
Thermo Scientific Chemicals

Acetate Buffer (1 M, pH 5.0, Low Endotoxin)

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J67248.AK
also known as J67248-AK
Price (USD)
-
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
CommentAll components are dissolved in UV treated 18.2 megohm-cm ± 1 water, low endotoxin water, followed by pH adjustment and filtered through 0.22 micron filter
pH5.0 ± 0.15
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only