LY 294002, 98%
LY 294002, 98%
LY 294002, 98%
Thermo Scientific Chemicals

LY 294002, 98%

CAS: 154447-36-6 | C19H17NO3, C19H17NO3 | 307.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J67289.MB
also known as J67289-MB
Price (USD)
358.65
Special offer
Online exclusive
Ends: 28-Dec-2026
422.00
Save 63.35
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
358.65
Special offer
Online exclusive
Ends: 28-Dec-2026
422.00
Save 63.35
Each
Specifications
Assay98%
CAS154447-36-6
DescriptionWhite to pale yellow
Formsolid
Formula Weight307.35g/mol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C