CP-31398, 98%
CP-31398, 98%
CP-31398, 98%
Thermo Scientific Chemicals

CP-31398, 98%

CAS: 259199-65-0 | C22H26N4O, C22H26N4O.2HCl | 362.48 g/mol
Have Questions?
Catalog NumberQuantity
J67317.MA
also known as J67317-MA
10 mg
Catalog number J67317.MA
also known as J67317-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
Assay98%
CAS259199-65-0
DescriptionYellow
FormSolid
Formula Weight453.39
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C