TOFA, 99%
TOFA, 99%
TOFA, 99%
Thermo Scientific Chemicals

TOFA, 99%

CAS: 54857-86-2 | C19H32O4, C19H32O4 | 324.46 g/mol
Have Questions?
Catalog NumberQuantity
J67337.MA
also known as J67337-MA
10 mg
Catalog number J67337.MA
also known as J67337-MA
Price (USD)
148.65
Online exclusive
165.00
Save 16.35
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
148.65
Online exclusive
165.00
Save 16.35
Each
Specifications
Assay99%
CAS54857-86-2
DescriptionOff-white
FormSolid
Formula Weight324.46
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C