NOB, 98%
NOB, 98%
NOB, 98%
Thermo Scientific Chemicals

NOB, 98%

CAS: 55894-52-5 | C15H19NO6, C15H19NO6 | 309.32 g/mol
Have Questions?
Catalog NumberQuantity
J67360.MF
also known as J67360-MF
50 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67360.MF
also known as J67360-MF
Price (USD)
-
Quantity:
50 mg
Request bulk or custom format
Specifications
Assay98%
CAS55894-52-5
DescriptionOff-white
FormSolid
Formula Weight309.32
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only