SQ 22536, 95%
SQ 22536, 95%
SQ 22536, 95%
Thermo Scientific Chemicals

SQ 22536, 95%

Have Questions?
Catalog NumberQuantity
J67455.LB0
also known as J67455-#0
5 mg
Catalog number J67455.LB0
also known as J67455-#0
Price (USD)
-
Quantity:
5 mg
Request bulk or custom format
Specifications
Assay95%
CAS17318-31-9
DescriptionWhite to off-white
FormSolid
Formula Weight205.22
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C