What are the concentrations of the components of Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol?
The Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol contains:
- 121 mM sodium chloride
- 1.5 mM calcium chloride
- 4.9 mM potassium chloride
- 1.0 mM magnesium chloride
- 0.3 mM magnesium sulfate
- 27 mM sodium bicarbonate
- 0.2 mM monopotassium phosphate
- 0.8 mM sodium phosphate dibasic
- 5.6 mM glucose