Product Image
Thermo Scientific Chemicals
Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J67569.AP
also known as J67569-AP
Price (USD)
93.80
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
93.80
Each
Specifications
ColorColorless
Recommended StorageAmbient Temperatures
Physical FormLiquid
Quantity500 mL
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What are the concentrations of the components of Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol?
The Gey's Balanced Salt Solution, with calcium, with magnesium, without phenol contains:
  • 121 mM sodium chloride
  • 1.5 mM calcium chloride
  • 4.9 mM potassium chloride
  • 1.0 mM magnesium chloride
  • 0.3 mM magnesium sulfate
  • 27 mM sodium bicarbonate
  • 0.2 mM monopotassium phosphate
  • 0.8 mM sodium phosphate dibasic
  • 5.6 mM glucose