Is the Krebs-Ringer Solution, bicarbonate-buffered sterile?
The Krebs-Ringer Solution, bicarbonate-buffered is prepared in 18.2 megohms water and filtered through 0.22 micron filter to remove contaminants. No sterility testing is performed so it cannot be certified as sterile.
What are the concentrations of the components of Krebs-Ringer Solution, bicarbonate-buffered?
The Krebs-Ringer Solution, bicarbonate-buffered contains 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5 mM D-glucose.