What is the pH of the Krebs-Ringer Solution, without calcium?
The pH of the Krebs-Ringer Solution, without calcium is 7.3.
Search
View additional product information for Krebs-Ringer Solution, without calcium - FAQs (J67839.K2, J67839.AP)
2 product FAQs found