Jasplakinolide
Invitrogen™

Jasplakinolide

Jasplakinolide is a macrocyclic peptide isolated from the marine sponge Jaspis johnstoni and is a potent inducer of actin polymerizationRead more
Have Questions?
Catalog NumberQuantity
J7473100 μg
Catalog number J7473
Price (USD)
679.65
Online exclusive
708.00
Save 28.35
Each
Quantity:
100 μg
Price (USD)
679.65
Online exclusive
708.00
Save 28.35
Each
Jasplakinolide is a macrocyclic peptide isolated from the marine sponge Jaspis johnstoni and is a potent inducer of actin polymerization in vitro by stimulating actin filament nucleation. Jasplakinolide competes with phalloidin for actin binding (Kd = 15 nM).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Molecular FormulaC36H45BrN4O6
Quantity100 μg
Recommended StorageStore in freezer (-5°C to -30°C)
Shipping ConditionRoom Temperature
Sub Cellular LocalizationActin, Cytoskeleton
Physical FormSolid
Product TypePeptide
Unit SizeEach

Citations & References (249)

Citations & References
Abstract
Actin filament cross-linking by MARCKS: characterization of two actin-binding sites within the phosphorylation site domain.
Authors:Yarmola EG, Edison AS, Lenox RH, Bubb MR
Journal:J Biol Chem
PubMed ID:11294839
We recently identified conformational changes that occur upon phosphorylation of myristoylated alanine-rich protein kinase C substrate (MARCKS) that preclude efficient cross-linking of actin filaments (Bubb, M. R., Lenox, R. H., and Edison, A. S. (1999) J. Biol. Chem. 274, 36472-36478). These results implied that the phosphorylation site domain of MARCKS ... More
Specification of actin filament function and molecular composition by tropomyosin isoforms.
Authors:Bryce NS, Schevzov G, Ferguson V, Percival JM, Lin JJ, Matsumura F, Bamburg JR, Jeffrey PL, Hardeman EC, Gunning P, Weinberger RP
Journal:Mol Biol Cell
PubMed ID:12631719
The specific functions of greater than 40 vertebrate nonmuscle tropomyosins (Tms) are poorly understood. In this article we have tested the ability of two Tm isoforms, TmBr3 and the human homologue of Tm5 (hTM5(NM1)), to regulate actin filament function. We found that these Tms can differentially alter actin filament organization, ... More
Caveolin-associated filamentous actin (Cav-actin) defines a novel F-actin structure in adipocytes.
Authors:Kanzaki M, Pessin JE
Journal:J Biol Chem
PubMed ID:12039946
Dynamic actin remodeling has been implicated in the translocation of the insulin-responsive glucose transporter 4 (GLUT4) to the plasma membrane in adipocytes. Here we show that fully differentiated 3T3L1 adipocytes have unique cortical filamentous actin structure, designated Cav-actin (caveolae-associated F-actin). During 3T3L1 adipocyte differentiation, rhodamine-phalloidin staining demonstrated the formation of ... More
Actin filament turnover removes bundles from Drosophila bristle cells.
Authors:Guild GM, Connelly PS, Vranich KA, Shaw MK, Tilney LG
Journal:J Cell Sci
PubMed ID:11861770
Drosophila bristle cells form enormous extensions that are supported by equally impressive scaffolds of modular, polarized and crosslinked actin filament bundles. As the cell matures and support is taken over by the secreted cuticle, the actin scaffold is completely removed. This removal begins during cell elongation and proceeds via an ... More
Gelsolin mediates collagen phagocytosis through a rac-dependent step.
Authors:Arora PD, Glogauer M, Kapus A, Kwiatkowski DJ, McCulloch CA
Journal:Mol Biol Cell
PubMed ID:14617805
The role of gelsolin, a calcium-dependent actin-severing protein, in mediating collagen phagocytosis, is not defined. We examined alpha 2 beta 1 integrin-mediated phagocytosis in fibroblasts from wild-type (WT) and gelsolin knockout (Gsn(-)) mice. After initial contact with collagen beads, collagen binding and internalization were 60% lower in Gsn(-) than WT ... More