What are the concentrations of the components in TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure?
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure contains 500 mM Tris, 60 mM KCl, and 2.8 M NaCl in high purity RO water.
Search
View additional product information for TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure - FAQs (J75892.K2, J75892.AE, J75892.K8)
1 product FAQs found