Recombinant Proteinase K, Ready-to-Use solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Recombinant Proteinase K, Ready-to-Use solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number J80002.AA
also known as J80002-AA
Price (USD)
230.65
Special offer
Online exclusive
Ends: 30-Jun-2026
271.00
Save 40.35 (15%)
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
230.65
Special offer
Online exclusive
Ends: 30-Jun-2026
271.00
Save 40.35 (15%)
Each
Chemical Identifiers
CAS39450-01-6
SpecificationsSpecification SheetSpecification Sheet
CommentSupplied in 10mM Tris-HCl, 1mM calcium acetate, pH 7.5 containing 50% glycerol.
CommentUnits/ml: minimum 400u/ml
Concentration20mg/ml
RNaseNone detected
DNaseNone detected
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.