GeneJET Whole Blood Genomic DNA Purification Mini Kit, 250 Preps - FAQs
GeneJET Whole Blood Genomic DNA Purification Mini Kit, 250 Preps - FAQs
View additional product information for GeneJET Whole Blood Genomic DNA Purification Mini Kit - FAQs (K0782, K0781)
5 product FAQs found
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Are MagJET and GeneJET blood nucleic acid purification kits compatible with animal blood?
Yes, the kits are compatible with animal blood for DNA/RNA purification.
Can I use a vacuum manifold with GeneJET kits?
The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.
At what temperature should I perform centrifugation steps in the GeneJET kits?
We recommend performing centrifugation steps at room temperature when the temperature is not specified.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.