CCF2-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF2 substrate which allows it to readily enter cells.Read more
Have Questions?
Catalog Number
Quantity
K1023
5 mg
Catalog number K1023
Price (USD)
29,530.00
Each
Quantity:
5 mg
Price (USD)
29,530.00
Each
CCF2-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF2 substrate which allows it to readily enter cells. Upon entry, cleavage by endogenous cytoplasmic esterases rapidly converts CCF2-AM into its negatively charged form, CCF2 or CCF4, which is retained in the cytosol. CCF2 is a Fluorescence Resonance Energy Transfer (FRET) substrate which consists of a cephalosporin core linking 7-hydroxycoumarin to fluorescein. This product is to be used as a FRET-based substrate for β-lactamase as a sensitive reporter of mammalian gene expression in the GeneBLAzer® technology.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Permeant
For Use With (Equipment)Flow Cytometer, Microplate Reader
Product LineGeneBLAzer
Quantity5 mg
Shipping ConditionRoom Temperature
SubstrateCCF2-AM
Detection MethodFluorescence
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Lactamase Substrate
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Frequently asked questions (FAQs)
Do you offer an alternative to GeneBLAzer In Vivo Detection Kit (Cat. No. 12578134)?
The following are two alternatives to the GeneBLAzer In Vivo Detection Kit:
- LiveBLAzer FRET-B/G Loading Kit with CCF2-AM (Cat. No. K1032, K1025)
- LiveBLAzer FRET-B/G Loading Kit with CCF4-AM (Cat. No. K1095, K1096, K1030)
Concerning CCF2-AM and CCF4-AM, these molecules differ by two carbons in the bridge that links the coumarin moiety to the lactam ring. Both are in the membrane-permeable, esterified forms, and can be used for assays in intact cells. CCF4-AM has better solubility properties than CCF2-AM and is thus best suited for screening applications. In addition, CCF4-AM has slightly better FRET and thus slightly lower background than CCF2-AM.
Note: We also offer the loading solutions as a separate kit, Loading Solutions (Cat. No. K1026), and CCF2-AM is also available as a standalone product (Cat. No. K1023).
Citations & References (1)
Citations & References
Abstract
Folate receptor alpha and caveolae are not required for Ebola virus glycoprotein-mediated viral infection.
Folate receptor alpha (FRalpha) has been described as a factor involved in mediating Ebola virus entry into cells (6). Furthermore, it was suggested that interaction with FRalpha results in internalization and subsequent viral ingress into the cytoplasm via caveolae (9). Descriptions of cellular receptors for Ebola virus and its entry ... More