Product Image
TR40 Concentrate
TR40 Concentrate
Have Questions?
Catalog NumberQuantity
K10311 L
Catalog number K1031
Price (USD)
4,115.00
Each
Quantity:
1 L
Price (USD)
4,115.00
Each
TR40 Concentrate is the suppression dye component of Solution C in the Loading Kits for CCF2-AM and CCF4-AM.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageStore at room temperature.
For Use With (Equipment)CCF2-AM and CCF4-AM
Product LineGeneBLAzer
Quantity1 L
Unit SizeEach