Solution D
Actual product may vary
Solution D
Solution D
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K115725 mL
K11561 mL
Catalog number K1157
Price (USD)
2,275.00
Each
Quantity:
25 mL
Price (USD)
2,275.00
Each
Solution D is an anion transport inhibitor, for cells such as CHO-K1, that can be used in conjunction with CCF2-AM and CCF4-AM-based studies to prevent active cellular export of the FRET-based substrates after loading.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageStore in freezer (-5 to -30°C).
For Use With (Application)Pharma & Biopharma; Protein Assays and Analysis; Cellular Imaging
Product LineGeneBLAzer
Quantity25 mL
Unit SizeEach