Rapid DNA Ligation Kit, 150 Reactions - FAQs
Rapid DNA Ligation Kit, 150 Reactions - FAQs
View additional product information for Rapid DNA Ligation Kit - FAQs (K1423, K1422)
1 product FAQs found
What is the difference between Rapid DNA Ligation Kit and Thermo Scientific T4 DNA ligase with a standard buffer?
The speed of ligation is the main difference between the two. The Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature. The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation.