Easy-DNA™ gDNA Purification Kit - FAQs
Easy-DNA™ gDNA Purification Kit - FAQs
View additional product information for Easy-DNA™ gDNA Purification Kit - FAQs (K180001)
1 product FAQs found
What type of samples can be used for the isolation of DNA using the Easy-DNA Kit?
DNA from the following sources has been isolated and successfully used to produce PCR products or in Southern blot experiments:
- Fresh, dried, frozen, or heparinized blood
- Tissue culture cells, both suspended and trypsinized
- Mammalian tissue
- E.coli
- Yeast cells
- Plant leaves
- Hair follicles
- Mouse tails
- Baculovirus (viral particles)