If I do not use all the wells in the filter plate of the PureLink Pro 96 Purification System, can I use those wells at another time?
Yes. Keep the unused wells covered with Foil Tape. Please note that extra Foil Tape and Receiver Plates will be needed.